APO455828400 Apollo Scientific Ltd

3-Allylphenol

  • MDL number MFCD20257851
  • Cas Number : 1446-24-8
  • Molecular Weight : 134.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Prop-2-en-1-yl)phenol
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 108-115/13mm
Product Link https://store.apolloscientific.co.uk/product/3-allylphenol

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-allylphenol
pictograms GHS05;GHS06
hstatements H301+H311;H314
Availability details for
Quantity
  • Availability
  • Add to Cart