APO455828411 Apollo Scientific Ltd

5-Amino-4-phenyl-1H-pyrazole

  • MDL number MFCD00231797
  • Cas Number : 5591-70-8
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 174-177
Iupac Name 4-phenyl-1H-pyrazol-5-ylamine
Inchi Code 1S/C9H9N3/c10-9-8(6-11-12-9)7-4-2-1-3-5-7/h1-6H,(H3,10,11,12)
Inchi Key QEHKQNYBBLCFIJ-UHFFFAOYSA-N

Description

Synonym 4-Phenyl-2H-pyrazol-3-amine, 4-Phenyl-1H-pyrazol-5-amine, 3-Amino-4-phenyl-1H-pyrazole, 4-Phenyl-1H-pyrazol-3-amine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 174-177
Product Link https://store.apolloscientific.co.uk/product/5-amino-4-phenyl-1h-pyrazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-4-phenyl-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart