APO455828418 Apollo Scientific Ltd

2-(1,3-Thiazol-2-yl)benzaldehyde

  • MDL number MFCD06657778
  • Cas Number : 223575-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(2-Formylphenyl)-1,3-thiazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40950

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40950
pictograms GHS06
hstatements H301;H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart