APO455828419 Apollo Scientific Ltd

2-Methoxy-6-[2-(pyrrolidin-1-yl)ethyl]aniline

  • MDL number MFCD00202972
  • Cas Number : 387358-44-3
  • Molecular Weight : 220.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-86
Synonym 1-[2-(2-Amino-3-methoxyphenyl)ethyl]pyrrolidine, 2-Amino-3-[2-(pyrrolidin-1-yl)ethyl]anisole
Purity 95
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methoxy-6-2-pyrrolidin-1-ylethylaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxy-6-2-pyrrolidin-1-ylethylaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart