APO455828427 Apollo Scientific Ltd

Ethyl 3,4-diaminobenzoate

  • MDL number MFCD00179337
  • Cas Number : 37466-90-3
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-113
Synonym 4-(Ethoxycarbonyl)benzene-1,2-diamine, 4-(Ethoxycarbonyl)phenylene-1,2-diamine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-34-diaminobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-34-diaminobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart