APO455828429 Apollo Scientific Ltd

Methyl 6-bromohexanoate

  • MDL number MFCD00187528
  • Cas Number : 14273-90-6
  • Molecular Weight : 209.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 6-bromohexanoate
density 1.316
Inchi Code 1S/C7H13BrO2/c1-10-7(9)5-3-2-4-6-8/h2-6H2,1H3
Inchi Key KYLVAMSNNZMHSX-UHFFFAOYSA-N

Description

Synonym Methyl 6-bromocaproate
Purity 98%
Density 1.316
Flash Point 70-74/1mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-6-bromohexanoate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-6-bromohexanoate
pictograms GHS07;GHS05
hstatements H314;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart