APO455828448 Apollo Scientific Ltd

3-Bromo-2-nitropyridine

  • MDL number MFCD00955614
  • Cas Number : 54231-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-106
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-nitropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-nitropyridine
pictograms GHS07
hstatements H302+H312;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart