APO455828459 Apollo Scientific Ltd

2,3,5,6-Tetrachloropyridine

  • MDL number MFCD00092428
  • Cas Number : 2402-79-1
  • Molecular Weight : 216.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-93
Purity 98
Flash Point 188
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 250-252
Product Link https://store.apolloscientific.co.uk/product/2356-tetrachloropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2356-tetrachloropyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart