APO455828461 Apollo Scientific Ltd

1-(4-Cyclohexylphenyl)-2-methylpropan-1-one

  • MDL number MFCD01098369
  • Cas Number : 53207-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4'-Cyclohexyl-2-methylpropiophenone, 4'-Cyclohexylisobutyrophenone
Purity 97
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32774

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32774
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart