APO455828464 Apollo Scientific Ltd

1-(2-Iodobenzyl)-1H-1,2,4-triazole

  • MDL number MFCD08741431
  • Cas Number : 876316-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67-69.5
Synonym 1-[(2-Iodophenyl)methyl]-1H-1,2,4-triazole, 1-Iodo-2-[(1H-1,2,4-triazol-1-yl)methyl]benzene
Purity 97
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32806

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32806
pictograms GHS02
hstatements H228
Availability details for
Quantity
  • Availability
  • Add to Cart