APO455828468 Apollo Scientific Ltd

1,2,3-Benzothiadiazole-5-carboxaldehyde

  • MDL number MFCD08690236
  • Cas Number : 394223-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-104
Synonym 5-Formyl-1,2,3-benzothiadiazole
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32779

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32779
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart