APO455828469 Apollo Scientific Ltd

1H-Benzimidazole-5-carboxaldehyde

  • MDL number MFCD08741423
  • Cas Number : 58442-17-4
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 159-161
Iupac Name 1H-benzimidazole-5-carbaldehyde
Inchi Code 1S/C8H6N2O/c11-4-6-1-2-7-8(3-6)10-5-9-7/h1-5H,(H,9,10)
Inchi Key ALCHVVTYAHQOFY-UHFFFAOYSA-N

Description

Synonym 5-Formyl-1H-benzimidazole
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 159-161
Product Link https://store.apolloscientific.co.uk/product/1h-benzimidazole-5-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1h-benzimidazole-5-carboxaldehyde
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart