APO455828487 Apollo Scientific Ltd

5-Methoxypyridine-3-boronic acid

  • MDL number MFCD03425977
  • Cas Number : 850991-69-4
  • Molecular Weight : 152.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224-227
Synonym 3-Borono-5-methoxypyridine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-methoxypyridine-3-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-methoxypyridine-3-boronic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart