APO455828498 Apollo Scientific Ltd

3,4-Dichlorocinnamic acid

  • MDL number MFCD00004385
  • Cas Number : 1202-39-7
  • Molecular Weight : 217.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-220
Synonym 3-(3,4-Dichlorophenyl)acrylic acid, 3-(3,4-Dichlorophenyl)prop-2-enoic acid
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/34-dichlorocinnamic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/34-dichlorocinnamic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart