APO455828505 Apollo Scientific Ltd

4-Chloro-2-nitro-1-(phenylsulphonyl)benzene

  • MDL number MFCD00052250
  • Cas Number : 86030-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121
Synonym 4-Chloro-2-nitrodiphenyl sulphone
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-chloro-2-nitro-1-phenylsulphonylbenzene

Safety Information

signalword Danger
pcodes P260;P280;P301+P330+P331
MSDS https://store.apolloscientific.co.uk/product/4-chloro-2-nitro-1-phenylsulphonylbenzene
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart