APO455828508 Apollo Scientific Ltd

2-Methoxy-6-methylphenol

  • MDL number MFCD00043771
  • Cas Number : 2896-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Hydroxy-3-methylanisole, 2-Hydroxy-3-methoxytoluene, 6-Methoxy-o-cresol
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-methoxy-6-methylphenol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxy-6-methylphenol
pictograms GHS07
hstatements H302;H319
Availability details for
Quantity
  • Availability
  • Add to Cart