APO455828510 Apollo Scientific Ltd

3-Chloropyridazine hydrochloride

  • MDL number MFCD12547058
  • Cas Number : 856847-77-3
  • Molecular Weight : 150.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloropyridazine hydrochloride
Inchi Code 1S/C4H3ClN2.ClH/c5-4-2-1-3-6-7-4;/h1-3H;1H
Inchi Key VYNOESFHQDZAMP-UHFFFAOYSA-N

Description

Synonym 3-Chloro-1,2-diazine hydrochloride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloropyridazine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloropyridazine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart