APO455828511 Apollo Scientific Ltd

Ethyl 4-amino-3,5-dichlorobenzoate

  • MDL number MFCD00016636
  • Cas Number : 74878-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,6-Dichloro-4-(ethoxycarbonyl)aniline
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=17467

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=17467
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart