APO455828513 Apollo Scientific Ltd

2-Aminocyclohexan-1-ol

  • MDL number MFCD00191368
  • Cas Number : 6850-38-0
  • Molecular Weight : 115.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-aminocyclohexanol
Inchi Code 1S/C6H13NO/c7-5-3-1-2-4-6(5)8/h5-6,8H,1-4,7H2
Inchi Key PQMCFTMVQORYJC-UHFFFAOYSA-N

Description

Synonym 1-Amino-2-hydroxycyclohexane, 2-Hydroxycyclohexan-1-amine
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-aminocyclohexan-1-ol

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-aminocyclohexan-1-ol
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart