APO455828515 Apollo Scientific Ltd

2,4-Bis(tert-butyl)-6-nitrophenol

  • MDL number MFCD00026282
  • Cas Number : 20039-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-65
Synonym 3,5-Bis(tert-butyl)-2-hydroxynitrobenzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=17496

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=17496
pictograms GHS07;GHS09
hstatements H302+H312+H332;H319;H413
Availability details for
Quantity
  • Availability
  • Add to Cart