APO455828527 Apollo Scientific Ltd

3-(4-Bromothien-2-yl)-1-(4-chlorophenyl)prop-2-en-1-one

  • MDL number MFCD00117349
  • Cas Number : 219689-82-4
  • Molecular Weight : 327.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-2-[3-(4-chlorophenyl)-3-oxoprop-1-en-1-yl]thiophene, 1-[(4-Bromothien-2-yl)acryloyl]-4-chlorobenzene
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-4-bromothien-2-yl-1-4-chlorophenylprop-2-en-1-one

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-4-bromothien-2-yl-1-4-chlorophenylprop-2-en-1-one
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart