APO455828531 Apollo Scientific Ltd

N'-Hydroxy-N-(5-methylisoxazol-3-yl)formamidine

  • MDL number MFCD00105523
  • Cas Number : 382136-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-187
Synonym N'-Hydroxy-N-(5-methyl-1,2-oxazol-3-yl)imidoformamide, 3-{[(Hydroxyimino)methyl]amino}-5-methylisoxazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=17603

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=17603
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart