APO455828532 Apollo Scientific Ltd

1-Acetamido-3-aminonaphthalene

  • MDL number MFCD09263886
  • Cas Number : 721970-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Acetylamino)-3-aminonaphthalene, N-(3-Aminonaphth-1-yl)acetamide
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=41538

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=41538
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart