APO455828534 Apollo Scientific Ltd

2-(Methylthio)nicotinoyl chloride

  • MDL number MFCD00051678
  • Cas Number : 97936-43-1
  • Molecular Weight : 187.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-96
Synonym 2-(Methylthio)pyridine-3-carbonyl chloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methylthionicotinoyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methylthionicotinoyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart