APO455828535 Apollo Scientific Ltd

Ethyl 7-azaindole-2-carboxylate

  • MDL number MFCD09998715
  • Cas Number : 221675-35-0
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Inchi Code 1S/C10H10N2O2/c1-2-14-10(13)8-6-7-4-3-5-11-9(7)12-8/h3-6H,2H2,1H3,(H,11,12)
Inchi Key CBIVEMGUNRNUEB-UHFFFAOYSA-N

Description

Synonym 2-(Ethoxycarbonyl)-1H-pyrrolo[2,3-b]pyridine, Ethyl 1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-7-azaindole-2-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-7-azaindole-2-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart