APO455828537 Apollo Scientific Ltd

Thieno[2,3-c]pyridine

  • MDL number MFCD01760492
  • Cas Number : 272-12-8
  • Molecular Weight : 135.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Azabenzo[b]thiophene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/thieno23-cpyridine

Safety Information

signalword Warning
pcodes P260;P280;P271
MSDS https://store.apolloscientific.co.uk/product/thieno23-cpyridine
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart