APO455828538 Apollo Scientific Ltd

2-(Ethylthio)nicotinoyl chloride

  • MDL number MFCD00051679
  • Cas Number : 123116-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-55(dec.)
Synonym 2-(Ethylthio)pyridine-3-carbonyl chloride
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=17628

Safety Information

signalword Danger
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=17628
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart