APO455828547 Apollo Scientific Ltd

3-(Piperidin-4-yl)-1H-indole

  • MDL number MFCD01764670
  • Cas Number : 17403-09-7
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 223-229
Iupac Name 3-(4-piperidinyl)-1H-indole
Inchi Code 1S/C13H16N2/c1-2-4-13-11(3-1)12(9-15-13)10-5-7-14-8-6-10/h1-4,9-10,14-15H,5-8H2
Inchi Key KAIRZPVWWIMPFT-UHFFFAOYSA-N

Description

Synonym 4-(1H-Indol-3-yl)piperidine
Purity 95%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 223-229
Product Link https://store.apolloscientific.co.uk/product/3-piperidin-4-yl-1h-indole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-piperidin-4-yl-1h-indole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart