APO455828548 Apollo Scientific Ltd

4-Bromo-2,3-dimethyl-6-nitroaniline

  • MDL number MFCD00051710
  • Cas Number : 108485-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-154
Synonym 2-Amino-5-bromo-3,4-dimethylnitrobenzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-23-dimethyl-6-nitroaniline

Safety Information

signalword Warning
pcodes P260;P280;P311
MSDS https://store.apolloscientific.co.uk/product/4-bromo-23-dimethyl-6-nitroaniline
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart