APO455828550 Apollo Scientific Ltd

3-(2-Aminothien-5-yl)pyridine

  • MDL number MFCD09038085
  • Cas Number : 837376-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126
Synonym 5-(Pyridin-3-yl)thien-2-amine, 2-Amino-5-(pyridin-3-yl)thiophene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=41718

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=41718
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart