APO455828553 Apollo Scientific Ltd

4,5-Dimethylpyridazine-3,6-diol

  • MDL number MFCD00067744
  • Cas Number : 5754-17-6
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-hydroxy-4,5-dimethyl-3(2H)-pyridazinone
Inchi Code 1S/C6H8N2O2/c1-3-4(2)6(10)8-7-5(3)9/h1-2H3,(H,7,9)(H,8,10)
Inchi Key QLFHLLQTPRTMLL-UHFFFAOYSA-N

Description

Synonym 3,6-Dihydroxy-4,5-dimethylpyridazine, 3,6-Dihydroxy-4,5-dimethyl-1,2-diazine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/45-dimethylpyridazine-36-diol

Safety Information

signalword Warning
pcodes P280;P260
MSDS https://store.apolloscientific.co.uk/product/45-dimethylpyridazine-36-diol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart