APO455828556 Apollo Scientific Ltd

1,6-Dihydro-6-oxopyridazine-3-carboxylic acid monohydrate

  • MDL number MFCD02090600
  • Cas Number : 306934-80-5
  • Molecular Weight : 158.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 246-247
Iupac Name 6-hydroxy-3-pyridazinecarboxylic acid hydrate
Inchi Code 1S/C5H4N2O3.H2O/c8-4-2-1-3(5(9)10)6-7-4;/h1-2H,(H,7,8)(H,9,10);1H2
Inchi Key DZCBVXKRERSHAA-UHFFFAOYSA-N

Description

Synonym 3-Carboxy-1,6-dihydro-6-oxopyridazine monohydrate
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 246-247
Product Link https://store.apolloscientific.co.uk/product/16-dihydro-6-oxopyridazine-3-carboxylic-acid-monohydrate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/16-dihydro-6-oxopyridazine-3-carboxylic-acid-monohydrate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart