APO455828557 Apollo Scientific Ltd

4-Amino-1-methyl-1H-indazole

  • MDL number MFCD09870041
  • Cas Number : 77894-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Methyl-1H-indazol-4-amine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-amino-1-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-1-methyl-1h-indazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart