APO455828559 Apollo Scientific Ltd

2-Bromo-5-chlorophenylacetic acid

  • MDL number MFCD00153890
  • Cas Number : 81682-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-103
Synonym (2-Bromo-5-chlorophenyl)acetic acid, 2-(2-Bromo-5-chlorophenyl)ethanoic acid
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-chlorophenylacetic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-chlorophenylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart