APO455828563 Apollo Scientific Ltd

4,5-Dibromo-2-phenyl-2,3-dihydropyridazin-3-one

  • MDL number MFCD00052907
  • Cas Number : 14305-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/45-dibromo-2-phenyl-23-dihydropyridazin-3-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/45-dibromo-2-phenyl-23-dihydropyridazin-3-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart