APO455828575 Apollo Scientific Ltd

2,4-Dibromobenzoic acid

  • MDL number MFCD00234253
  • Cas Number : 611-00-7
  • Molecular Weight : 279.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 50-52
Iupac Name 2,4-dibromobenzoic acid
Inchi Code 1S/C7H4Br2O2/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3H,(H,10,11)
Inchi Key NAGGYODWMPFKJQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 50-52
Product Link https://store.apolloscientific.co.uk/product/24-dibromobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-dibromobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart