APO455828576 Apollo Scientific Ltd

Methyl 3,3-bis(methylthio)-2-cyanoacrylate

  • MDL number MFCD00067841
  • Cas Number : 3490-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 3,3-bis(methylsulphanyl)-2-cyanoprop-2-enoate
Purity 95
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=17933

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=17933
Availability details for
Quantity
  • Availability
  • Add to Cart