APO455828580 Apollo Scientific Ltd

1-(3,4-Dichlorophenyl)biguanide hydrochloride

  • MDL number MFCD00173791
  • Cas Number : 21703-08-2
  • Molecular Weight : 282.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222-228
Synonym N-(3,4-Dichlorophenyl)imidodicarbonimidic diamide hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-34-dichlorophenylbiguanide-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-34-dichlorophenylbiguanide-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart