APO455828586 Apollo Scientific Ltd

3-Methoxyphenylacetic acid hydrazide

  • MDL number MFCD00085106
  • Cas Number : 34624-38-9
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 89-90
Iupac Name 2-(3-methoxyphenyl)acetohydrazide
Inchi Code 1S/C9H12N2O2/c1-13-8-4-2-3-7(5-8)6-9(12)11-10/h2-5H,6,10H2,1H3,(H,11,12)
Inchi Key XEALVGVRTCEVES-UHFFFAOYSA-N

Description

Synonym 2-(3-Methoxyphenyl)acetohydrazide
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 89-90
Product Link https://store.apolloscientific.co.uk/product/3-methoxyphenylacetic-acid-hydrazide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-methoxyphenylacetic-acid-hydrazide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart