APO455828588 Apollo Scientific Ltd

Methyl 4,5-dihydro-4-(2-methoxy-2-oxoethyl)-2-methyl-5-oxo-1H-pyrrole-3-carboxylate

  • MDL number MFCD00173858
  • Cas Number : 77978-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-94
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=18017

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=18017
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart