APO455828589 Apollo Scientific Ltd

Ethyl 3,5-dimethyl-4-ethyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD00030376
  • Cas Number : 2199-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-96
Synonym 3,5-Dimethyl-2-(ethoxycarbonyl)-4-ethyl-1H-pyrrole, 2,4-Dimethyl-5-(ethoxycarbonyl)-3-ethyl-1H-pyrrole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=18020

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=18020
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart