APO455828598 Apollo Scientific Ltd

3-Amino-2-(2,3-dimethylphenoxy)pyridine

  • MDL number MFCD00173657
  • Cas Number : 175136-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-145
Synonym 2-(2,3-Dimethylphenoxy)pyridin-3-amine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=18074

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=18074
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart