APO455828612 Apollo Scientific Ltd

4-(Oct-1-yl)bromobenzene

  • MDL number MFCD00038348
  • Cas Number : 51554-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(4-Bromophenyl)octane
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-oct-1-ylbromobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-oct-1-ylbromobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart