APO455828616 Apollo Scientific Ltd

4'-Iodoacetanilide

  • MDL number MFCD00016352
  • Cas Number : 622-50-4
  • Molecular Weight : 261.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(4-iodophenyl)acetamide
Inchi Code 1S/C8H8INO/c1-6(11)10-8-4-2-7(9)3-5-8/h2-5H,1H3,(H,10,11)
Inchi Key SIULLDWIXYYVCU-UHFFFAOYSA-N

Description

Synonym N-(4-Iodophenyl)acetamide
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-iodoacetanilide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-iodoacetanilide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart