APO455828618 Apollo Scientific Ltd

3,5-Bis(tert-butyl)-4-hydroxyphenylacetonitrile

  • MDL number MFCD00156139
  • Cas Number : 1611-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-112
Synonym [3,5-Bis-(tert-butyl)-4-hydroxyphenyl]acetonitrile, 3,5-Bis(tert-butyl)-4-hydroxybenzyl cyanide, 2,6-Bis(tert-butyl)-4-(cyanomethyl)phenol
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=18151

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=18151
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart