APO455828628 Apollo Scientific Ltd

Methyl 2-bromo-5-chlorobenzoate

  • MDL number MFCD00144763
  • Cas Number : 27007-53-0
  • Molecular Weight : 249.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 37-40
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-bromo-5-chlorobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-bromo-5-chlorobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart