APO455828629 Apollo Scientific Ltd

2-Allyl-3-hydroxybenzaldehyde

  • MDL number MFCD00100646
  • Cas Number : 79950-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-98
Synonym 3-Hydroxy-2-(prop-2-en-1-yl)benzaldehyde, 2-Allyl-3-formylphenol
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-allyl-3-hydroxybenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-allyl-3-hydroxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart