APO455828630 Apollo Scientific Ltd

Methyl 3-chloro-5-iodobenzoate

  • MDL number MFCD00673018
  • Cas Number : 289039-85-6
  • Molecular Weight : 296.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 41-43
Iupac Name methyl 3-chloro-5-iodobenzoate
Inchi Code 1S/C8H6ClIO2/c1-12-8(11)5-2-6(9)4-7(10)3-5/h2-4H,1H3
Inchi Key ZDSIXLJPASGHDW-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 41-43
Product Link https://store.apolloscientific.co.uk/product/methyl-3-chloro-5-iodobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-chloro-5-iodobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart