APO455828634 Apollo Scientific Ltd

4'-Amino-2'-hydroxyacetophenone

  • MDL number MFCD00100636
  • Cas Number : 2476-29-1
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-131
Synonym 1-(4-Amino-2-hydroxyphenyl)ethan-1-one, 4-Acetyl-3-hydroxyaniline, 2-Acetyl-5-aminophenol
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-2-hydroxyacetophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-2-hydroxyacetophenone
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart