APO455828635 Apollo Scientific Ltd

3'-Chloro-2'-hydroxyacetophenone

  • MDL number MFCD00100492
  • Cas Number : 3226-34-4
  • Molecular Weight : 170.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 47-49
Iupac Name 1-(3-chloro-2-hydroxyphenyl)ethanone
Inchi Code 1S/C8H7ClO2/c1-5(10)6-3-2-4-7(9)8(6)11/h2-4,11H,1H3
Inchi Key GBWVDQBTXFIIJF-UHFFFAOYSA-N

Description

Synonym 1-(3-Chloro-2-hydroxyphenyl)ethan-1-one, 2-Acetyl-6-chlorophenol
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Boiling point 47-49
Product Link https://store.apolloscientific.co.uk/product/3-chloro-2-hydroxyacetophenone

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/3-chloro-2-hydroxyacetophenone
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart